Potassium chloride ≥99.5% (by argentometric titration), Puriss. p.a.

Supplier: Honeywell Research Chemicals

60130-1KG 60130-250G 60130-5KG
BJ60130-1KGEA 171.69 USD
BJ60130-1KG BJ60130-250G BJ60130-5KG
Potassium chloride ≥99.5% (by argentometric titration), Puriss. p.a.
Potassium chloride
Formula: KCl
Boiling Pt: 1.413 °C (1.013 hPa)
Melting Pt: 772 °C
Density: 2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
EINECS: 231-211-8

Order Now

Specification Test Results

Assay (argentometric) 99.5 - 101.0 %
pH (5 %, 20°C) 5.5 - 8.0
Calcium (Ca) Max. 0.005 %
Cadmium (Cd) Max. 0.0005 %
Cobalt (Co) Max. 0.0005 %
Chromium (Cr) Max. 0.0005 %
Copper (Cu) Max. 0.0005 %
Iron (Fe) Max. 0.0002 %
Magnesium (Mg) Max. 0.001 %
Manganese (Mn) Max. 0.0005 %
Sodium (Na) Max. 0.02 %
Nickel (Ni) Max. 0.0005 %
Lead (Pb) Max. 0.0005 %
Zinc (Zn) Max. 0.0005 %
Bromide (Br) Max. 0.05 %
Iodide (I) Max. 0.002 %
Phosphate (PO₄) Max. 0.0005 %
Sulfate (SO₄) Max. 0.003 %
Total N Max. 0.001 %
Colour of the solution complying
Turbidity of solution complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR