Potassium carbonate, anhydrous ≥99.0% (by titrimetric analysis), Purum p.a.

Supplier: Honeywell Research Chemicals
Warning

60110-1KG
BJ60110-1KGEA 160.57 USD
BJ60110-1KG BJ60110-5KG
Potassium carbonate, anhydrous ≥99.0% (by titrimetric analysis), Purum p.a.
Potassium carbonate

  • Products with a guarantee certificate and/or suitable for the stated analytical application.

Formula: K₂CO₃
Boiling Pt: Decomposition
Melting Pt: 891 °C
Density: 2.430 g/cm³ (19 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011382
CAS Number: 584-08-7
EINECS: 209-529-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d5900faa1ba52c3213eed8f6bb2d952bac1f30f&vid=6298361525e6af416556b5694c74a8a91faa437c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.0 - 101.0 %
Calcium (Ca) Max. 50 ppm
Cadmium (Cd) Max. 50 ppm
Cobalt (Co) Max. 50 ppm
Copper (Cu) Max. 50 ppm
Iron (Fe) Max. 50 ppm
Sodium (Na) Max. 5000 ppm
Nickel (Ni) Max. 50 ppm
Lead (Pb) Max. 50 ppm
Zinc (Zn) Max. 50 ppm
Total S (as SO₄) Max. 50 ppm
Chloride (Cl) Max. 50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR