Water with 0,1% (v/v) formic acid 0.1% (v/v) in water, CHROMASOLV™, Solvent Blend for LC-MS, Riedel-de Haën™

Supplier: Honeywell Research Chemicals

576913-4X4L
BJ576913-4X4LCS 821 USD
BJ576913-4X4L
Water with 0,1% (v/v) formic acid 0.1% (v/v) in water, CHROMASOLV™, Solvent Blend for LC-MS, Riedel-de Haën™
Water with 0,1% (v/v) formic acid

  • High purity chromatography solvents
  • Suitability tested for specified use from HPLC up to UHPLC-MS applications.


UNSPSC Code: 12190000

Density: 0.998 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 64-18-6
EINECS: 231-791-2

Order Now

Specification Test Results

Contains 0.1 %(v/v) formic acid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR