Ammonium formate ≥99% (by anhydrous basis) eluent additive, for LC-MS, Fluka™

Supplier: Honeywell Research Chemicals
Warning

Synonyms: Formic acid ammonium salt

55674-50G
BJ55674-50GEA 550 USD
BJ55674-50G
Ammonium formate ≥99% (by anhydrous basis) eluent additive, for LC-MS, Fluka™
Ammonium formate
Formula: HCO₂NH₄
MW: 63.06 g/mol
Melting Pt: 116 °C
Density: 1.266 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013103
CAS Number: 540-69-2
EINECS: 208-753-9

Order Now

Specification Test Results

Assay (calculated on the anhydrous basis) 99.0 - 101.0 %
Water (Karl Fischer) Max. 2 %
Sulfated ash Max. 0.1 %
Aluminium (Al) Max. 1 ppm
Arsenic (As) Max. 0.1 ppm
Barium (Ba) Max. 1 ppm
Bismuth (Bi) Max. 5 ppm
Calcium (Ca) Max. 5 ppm
Cadmium (Cd) Max. 1 ppm
Cobalt (Co) Max. 1 ppm
Chromium (Cr) Max. 1 ppm
Copper (Cu) Max. 1 ppm
Iron (Fe) Max. 1 ppm
Potassium (K) Max. 5 ppm
Lithium (Li) Max. 5 ppm
Magnesium (Mg) Max. 1 ppm
Manganese (Mn) Max. 1 ppm
Molybdenum (Mo) Max. 5 ppm
Sodium (Na) Max. 5 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Strontium (Sr) Max. 1 ppm
Zinc (Zn) Max. 1 ppm
Chloride (Cl) Max. 50 ppm
Total S (as SO₄) Max. 50 ppm
Suitability for the LC-MS complying
Appearance of the solution complying
Solution 1 M in H₂O complying

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR