1-Butanol ≥99.9%

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Propylcarbinol, Methylolpropane, Butan-1-ol, Butyl alcohol, n-Butyl alcohol, Butyl hydroxide, 1-Butyl alcohol, Butyl Alcohol, 1-Hydroxybutane, 1-Hydroxy butane, n-Butan-1-ol, Propyl carbinol, n-Butanol, n-Butyl Alcohol

537993-20L 537993-1L 537993-2.5L 537993-6X1L 537993-4X2.5L
BJ537993-20LEA 1333.4 USD
BJ537993-20L BJ537993-1L BJ537993-2.5L BJ537993-6X1L BJ5379934X2.5L
1-Butanol ≥99.9%
1-Butanol

We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:


  • Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
  • Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.


Beilstein Number: 969148
RTECS Number: EO1400000
UNSPSC Code: 12191601

Formula: H₃C(CH₂)₂CH₂OH
MW: 74.12 g/mol
Boiling Pt: 117.6
Melting Pt: -89
Density: 0.81 g/cm³ (20 °C)
Flash Pt: 9.4 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00002964
CAS Number: 71-36-3
EINECS: 200-751-6
UN: 1120
ADR: 3 III
Merck Index: 13,01539
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9395eeb6150a74222da52460f53cedeccd93442&vid=fd8352a556914c5be7213c576355a88e130d1a7b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity (IR) complying
Assay (GC) Min. 99.80 %
Density (D 20/4) 0.809 - 0.811
Non-volatile matter Max. 20 mg/l
Water (Karl Fischer) Max. 0.1 %
Free acid (as CH₃COOH) Max. 0.005 %
Iron (Fe) Max. 0.3 ppm
Di-n-butyl ether (GC) Max. 0.100 %
Isobutyl alcohol (GC) Max. 0.100 %
n-Butyraldehyde (GC) Max. 0.020 %
Carbonyl compounds (as CO) Max. 0.050 %
APHA Max. 10
Autoignition Temperature 649 °F
Refractive Index n20/D 1.399 (lit.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR