Xylene (mixture of isomers) for histology

Supplier: Honeywell Research Chemicals
Danger

534056-4L
BJ534056-4LEA 204 USD
BJ534056-4L
Xylene (mixture of isomers) for histology
Xylene (mixture of isomers)

  • "Commonly used for tissue sampling and processing
  • Precise control of the manufacturing process ensures unsurpassed lot-to-lot consistency
  • Available in a variety of packaging options (amber glass bottles, carbon steel flex-spout cans and carbon steel cans with NPT threads)"

Formula: C₆H₄(CH₃)₂
Boiling Pt: 136 - 140 °C
Melting Pt: -34 °C
Density: 0.87 g/cm³
Flash Pt: 25 °C
Storage Temperature: Ambient
MDL Number: MFCD00077264
CAS Number: 1330-20-7
UN: 1307
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2deefc7b7b67c10e64003e243ff3216fad27cf7d&vid=9fa01ddafd4814d5f34266ae7933463a29d92d19&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Vapor Pressure 18 mmHg ( 37.7 °C)
IR spectrum complying
C₈ isomers including Ethylbenzene (GC) Min. 96 %
Total isomers (GC) Min. 73 %
Non-volatile matter Max. 0.002 %
Water (Karl Fischer) Max. 0.05 %
Ethyl benzene (GC) Max. 25.0 %
APHA Max. 10


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR