N,N-Dimethylformamide ≥99.9%, CHROMASOLV™ for headspace gas chromatography, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

51781-6X1L
BJ51781-1LEA 690.97 USD
BJ51781-1L BJ51781-6X1L
N,N-Dimethylformamide ≥99.9%, CHROMASOLV™ for headspace gas chromatography, Riedel-de Haën™
N,N-Dimethylformamide

Honeywell Chromasolv™ GC-headspace solvents:


  • Developed particularly for the identification and quantification of residual solvents in pharmaceuticals using GC-headspace methods described in monographs from the United States Pharmacopeia (USP) or the European Pharmacopoeia (Ph. Eur.) and following the International Conference on Harmonization of Technical Requirements for Registration of Pharmaceuticals for Human Use (ICH) guidelines
  • Very pure solvents with extremely low concentrations of the defined residual solvents

Formula: HCON(CH₃)₂
Boiling Pt: 152…153 °C (1.013 hPa)
Melting Pt: –61 °C
Density: 0.94 g/cm³ (20 °C)
Flash Pt: 57.5 °C (1.013.25 hPa)
Storage Temperature: Ambient
MDL Number: MFCD00003284
CAS Number: 68-12-2
EINECS: 200-679-5
UN: 2265
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ac5c5e9758b6c40ccb0cf127b6a6e1c6d608c64&vid=9a5d4e579e195193602c20d3cb39c89ac9c92cbf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Suitability for the GC Headspace complying
Assay (GC) Min. 99.9 %
Refractive index (n 20/D) 1.429 - 1.431
Water (Karl Fischer) Max. 0.03 %
Absorbance at 270 nm Max. 0.700
Absorbance at 275 nm Max. 0.250
Absorbance at 300 nm Max. 0.070
Absorbance at 320 nm Max. 0.022


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR