Pumice stone, granular 4-6 mm, Fluka™

Supplier: Honeywell Research Chemicals

51753-1KG
BJ51753-1KGEA 135 USD
BJ51753-1KG
Pumice stone, granular 4-6 mm, Fluka™
Pumice stone
Storage Temperature: Ambient
MDL Number: MFCD00148147
CAS Number: 1332-09-8

Order Now

Specification Test Results

Grain Size 5-8 Mm Min. 85 %
Loss on ignition (800°C) Max. 4 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR