N-Methyl-2-pyrrolidone (NMP) ≥99.7%, Biotechnology Grade, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: 1-Methyl-2-pyrrolidinone, N-Methyl-2-pyrrolidinone,NMP, N-Methylpyrrolidinone, N-Methyl-2-pyrrolidinone, N-Methyl-alpha-pyrrolidone, M-Pyrol, N-methylpyrrolidone, N-Methyl-gamma-butyrolactam, 1-Methyl-2-pyrrolidone, NMP, N-Methylpyrrolidone, N-Methyl-2-pyrrolidone, Methyl Pyrrolidone

494496-1L 494496-2.5L
BJ494496-1LEA 265.98 USD
BJ494496-1L BJ494496-2.5L BJ494496-2L
N-Methyl-2-pyrrolidone (NMP) ≥99.7%, Biotechnology Grade, Burdick & Jackson™
N-Methyl-2-pyrrolidone (NMP)
Formula: C₅H₉NO
MW: 99.13 g/mol
Boiling Pt: 202
Melting Pt: -24
Density: 1.028 g/cm³ (20 °C)
Flash Pt: 9.5 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00003193
CAS Number: 872-50-4
EINECS: 212-828-1
Merck Index: 13,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32d156c242c81ef2974339109ced3895beda87fd&vid=06c87bbb3b5050362f47a8fc836cedc8a6d51530&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) Min. 99.7 %
Water (Karl Fischer) Max. 0.005 %
Titrable Base Max. 0.001 meq/g
Free alkali (as methylamine) Max. 0.01 %
Absorbance at 400 nm Max. 0.01
Absorbance at 350 nm Max. 0.01
Absorbance at 325 nm Max. 0.10
Absorbance at 300 nm Max. 0.50
Absorbance at 285 nm Max. 1.00
APHA Max. 20


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR