Hydrofluoric acid 47 - 51%, TraceSELECT™ for trace analysis, Fluka™

Supplier: Honeywell Research Chemicals
Danger

47559-500ML
BJ47559-500MLEA 158 USD
BJ47559-500ML BJ47559-100ML
Hydrofluoric acid 47 - 51%, TraceSELECT™ for trace analysis, Fluka™
Hydrofluoric acid
Formula: HF
Boiling Pt: 105 °C (1.013 hPa)
Melting Pt: –35 °C
Density: 1.170 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011346
CAS Number: 7664-39-3
UN: 1790
ADR: 8,II

Order Now

Specification Test Results

Assay 47.0 - 51.0 %
Density (D 20/4) 1.17
Silver (Ag) Max. 0.001 ppm
Aluminium (Al) Max. 0.005 ppm
Arsenic (As) Max. 0.005 ppm
Gold (Au) Max. 0.001 ppm
Barium (Ba) Max. 0.001 ppm
Beryllium (Be) Max. 0.001 ppm
Bismuth (Bi) Max. 0.001 ppm
Calcium (Ca) Max. 0.01 ppm
Cadmium (Cd) Max. 0.001 ppm
Cesium (Cs) Max. 0.001 ppm
Cobalt (Co) Max. 0.001 ppm
Chromium (Cr) Max. 0.001 ppm
Copper (Cu) Max. 0.001 ppm
Iron (Fe) Max. 0.01 ppm
Gallium (Ga) Max. 0.001 ppm
Germanium (Ge) Max. 0.001 ppm
Mercury (Hg) Max. 0.005 ppm
Indium (In) Max. 0.001 ppm
Potassium (K) Max. 0.01 ppm
Lithium (Li) Max. 0.001 ppm
Magnesium (Mg) Max. 0.005 ppm
Manganese (Mn) Max. 0.001 ppm
Molybdenum (Mo) Max. 0.001 ppm
Sodium (Na) Max. 0.01 ppm
Nickel (Ni) Max. 0.001 ppm
Lead (Pb) Max. 0.002 ppm
Rubidium (Rb) Max. 0.001 ppm
Selenium (Se) Max. 0.01 ppm
Tin (Sn) Max. 0.001 ppm
Strontium (Sr) Max. 0.001 ppm
Titanium (Ti) Max. 0.005 ppm
Thallium (Tl) Max. 0.001 ppm
Vanadium (V) Max. 0.001 ppm
Zinc (Zn) Max. 0.002 ppm
Zirconium (Zr) Max. 0.001 ppm
Vapor Density 1.27 (vs air)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR