ACETIC ACID 0.1% /METH 4L
Supplier: Honeywell Research Chemicals
0.1% Acetic Acid in Methanol (v/v). A high purity solution suitable for analytical applications such as HPLC. Packaged in 4-liter glass bottles, with 4 bottles per case.