Acetic acid ≥98%, tested according to Ph. Eur., Fluka™

Supplier: Honeywell Research Chemicals
Danger

45741-1L
BJ45741-1LEA 104.31 USD
BJ45741-1L
Acetic acid ≥98%, tested according to Ph. Eur., Fluka™
Acetic acid
Formula: H₃CCOOH
Boiling Pt: 118 °C (1.013 hPa)
Melting Pt: 16 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 40 °C
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Explosion Limit 16 %, 92 °F
Identity A complying
Identity B complying
Appearance of the substance complying
Assay 99.0 - 100.5 %
Non-volatile matter Max. 0.01 %
Congealing - Freezing Point Min. 14.8 °C
Iron (Fe) Max. 5 ppm
Heavy metals (as Pb) Max. 5 ppm
Chloride (Cl) Max. 23 ppm
Sulfate (SO₄) Max. 47 ppm
Reducing impurities complying
Residual solvents complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR