Calcon® (mordant black 17) indicator for complexometry (Al, Fe, Zr), Fluka™

Supplier: Honeywell Research Chemicals
Warning

45550-25G
BJ45550-100GEA 56.57 USD
BJ45550-100G BJ45550-25G
Calcon® (mordant black 17) indicator for complexometry (Al, Fe, Zr), Fluka™
Calcon® (Mordant Black 17)
Formula: C₂₀H₁₃N₂NaO₅S
Storage Temperature: Ambient
MDL Number: MFCD00004069
CAS Number: 2538-85-4
EINECS: 219-810-2

Order Now

Specification Test Results

Indicator solution complying
Suitability for Complexometry complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR