1-Dodecanol ≥98.0% ACS, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Warning

443816-1KG
BJ443816-1KGEA 206.33 USD
BJ443816-1KG BJ443816-500G
1-Dodecanol ≥98.0% ACS, Riedel-de Haën™
1-Dodecanol

ACS GENERAL USE SOLVENTS


  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Formula: H₃C(CH₂)₁₀CH₂OH
Boiling Pt: 260…262 °C
Melting Pt: 22…26 °C
Flash Pt: 121 °C
Storage Temperature: Ambient
MDL Number: MFCD00004753
CAS Number: 112-53-8
EINECS: 203-982-0

Order Now

Specification Test Results

Identity (IR) complying
Assay (GC) Min. 98.0 %
Water (Karl Fischer) Max. 0.05 %
APHA Max. 10
Vapor Density 7.4 (vs air)
Explosion Limit 4 %
Autoignition Temperature 500 °F
Vapor Pressure 0.1 mmHg ( 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR