Magnesium 2% in nitric acid 5% ≥99.9995%, TraceSELECT™ matrix modifier solution, Fluka™

Supplier: Honeywell Research Chemicals
Danger

428892-100ML
BJ428892-100MLEA 316 USD
BJ428892-100ML
Magnesium 2% in nitric acid 5% ≥99.9995%, TraceSELECT™ matrix modifier solution, Fluka™
Magnesium

The acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (µg/kg) trace range.


Packaged in high-quality containers appropriate for each product
the reagents are produced and bottled under clean-room conditions.

Formula: Mg
MW: 24.3 g/mol
Melting Pt: no data available
Density: 1.093 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085308
CAS Number: 7439-95-4
UN: 3264
ADR: 8,III

Order Now

Specification Test Results

Assay of Mg 1.5 - 2.5 %
Nitric acid 1.0 - 5.0 %
Metal trace analysis (ICP) Max. 5.5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR