Sodium hypochlorite 10 - 15% available chlorine in aqueous solution, Reagent Grade

Supplier: Honeywell Research Chemicals
Danger

425044-1L 425044-250ML
BJ425044-1LEA 174.9 USD
BJ425044-1L BJ425044-250ML
Sodium hypochlorite 10 - 15% available chlorine in aqueous solution, Reagent Grade
Sodium hypochlorite
Formula: ClNaO
Boiling Pt: 111 °C
Melting Pt: –30…–20 °C
Density: 1.206 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011120
CAS Number: 7681-52-9
UN: 1791
ADR: 8,II

Order Now

Specification Test Results

Assay of active chlorine 10 - 15 %
Composition available chlorine, 10-15%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR