1,1,1,3,3,3-Hexafluoro-2-propanol ≥99.8% eluent additive, for LC-MS, Fluka™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: 1,1,1,3,3,3-Hexafluoroisopropyl alcohol, Hexafluoro-2-propanol, Hexafluoroisopropanol, 1,1,1,3,3,3-Hexafluoropropan-2-ol, HFIP

42060-10X1ML
BJ42060-10X1MLPK 319 USD
BJ42060-10X1ML BJ42060-50ML
1,1,1,3,3,3-Hexafluoro-2-propanol ≥99.8% eluent additive, for LC-MS, Fluka™
1,1,1,3,3,3-Hexafluoro-2-propanol
Formula: C₃H₂F₆O
MW: 168.04 g/mol
Boiling Pt: 59 °C
Melting Pt: –4 °C
Density: 1.596 g/cm³ (25 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00011651
CAS Number: 920-66-1
EINECS: 213-059-4
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Assay (GC) Min. 99.8 %
Suitability for the LC-MS complying
Non-volatile matter Max. 5 ppm
Water (Karl Fischer) Max. 200 ppm
Absorbance at 240 nm Max. 0.01
Absorbance at 200 nm Max. 0.3
Aluminium (Al) Max. 0.5 ppm
Barium (Ba) Max. 0.1 ppm
Bismuth (Bi) Max. 0.1 ppm
Calcium (Ca) Max. 0.5 ppm
Cadmium (Cd) Max. 0.05 ppm
Cobalt (Co) Max. 0.02 ppm
Chromium (Cr) Max. 0.02 ppm
Copper (Cu) Max. 0.02 ppm
Iron (Fe) Max. 0.1 ppm
Potassium (K) Max. 0.5 ppm
Lithium (Li) Max. 0.1 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.02 ppm
Molybdenum (Mo) Max. 0.1 ppm
Sodium (Na) Max. 1 ppm
Nickel (Ni) Max. 0.02 ppm
Lead (Pb) Max. 0.1 ppm
Strontium (Sr) Max. 0.1 ppm
Zinc (Zn) Max. 0.1 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR