Methanol ≥99.9%, CHROMASOLV™ for GC/MS analysis of volatile organics, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

414816-6X1L
BJ414816-1LCS 1082.6 USD
BJ414816-1L BJ-414816-1L
Methanol ≥99.9%, CHROMASOLV™ for GC/MS analysis of volatile organics, Riedel-de Haën™
Methanol

Honeywell Chromasolv™ GC-headspace solvents:


  • developed particularly for the identification and quantification of residual solvents in pharmaceuticals using GC-headspace methods described in monographs from the United States Pharmacopeia (USP) or the European Pharmacopoeia (Ph. Eur.) and following the International Conference on Harmonization of Technical Requirements for Registration of Pharmaceuticals for Human Use (ICH) guidelines
  • very pure solvents with extremely low concentrations of the defined residual solvents
  • state-of-the art production and application tests


Beilstein Number: 1098229

Formula: CH₃OH
Boiling Pt: 64…65 °C (1.013 hPa)
Melting Pt: –98 °C
Density: 0.79 g/cm³ (20 °C)
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

IR spectrum complying
Assay (GC) min. 99.90 %
Non-volatile matter max. 1 ppm
Water (Karl Fischer) max. 0.2 %
Impurity Profile complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR