Acetone, Semiconductor grade VLSI (Honeywell 17617)

Supplier: Honeywell Research Chemicals
Danger

40289H-2.5L
BJ40289H-2.5LEA 231.55 USD
BJ40289H-2.5L
Acetone, Semiconductor grade VLSI (Honeywell 17617)
Acetone

We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:


  • Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
  • Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.


Beilstein Number: 635680

Formula: CH₃COCH₃
Boiling Pt: 55…57 °C (1.013 hPa)
Melting Pt: –95 °C
Density: 0.79 g/cm³ (20 °C)
Flash Pt: 14.3 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Particles > 0,5 µm Max. 250 P/ml
Assay (GC) Min. 99.5 %
Boiling range 55.5 - 56.5 °C
Density (D 20/4) 0.790 - 0.792
Refractive index (n 20/D) 1.3580 - 1.3600
Non-volatile matter Max. 5 ppm
Water (Karl Fischer) Max. 0.2 %
Free acid (as CH₃COOH) Max. 20 ppm
Free alkali (as NH₃) Max. 2 ppm
Silver (Ag) Max. 0.01 ppm
Aluminium (Al) Max. 0.05 ppm
Arsenic (As) Max. 0.01 ppm
Gold (Au) Max. 0.02 ppm
Boron (B) Max. 0.01 ppm
Barium (Ba) Max. 0.02 ppm
Beryllium (Be) Max. 0.01 ppm
Bismuth (Bi) Max. 0.02 ppm
Calcium (Ca) Max. 0.05 ppm
Cadmium (Cd) Max. 0.01 ppm
Cobalt (Co) Max. 0.01 ppm
Chromium (Cr) Max. 0.01 ppm
Copper (Cu) Max. 0.01 ppm
Iron (Fe) Max. 0.05 ppm
Gallium (Ga) Max. 0.01 ppm
Indium (In) Max. 0.01 ppm
Potassium (K) Max. 0.02 ppm
Lithium (Li) Max. 0.01 ppm
Magnesium (Mg) Max. 0.02 ppm
Manganese (Mn) Max. 0.01 ppm
Molybdenum (Mo) Max. 0.01 ppm
Sodium (Na) Max. 0.05 ppm
Nickel (Ni) Max. 0.01 ppm
Lead (Pb) Max. 0.01 ppm
Platinum (Pt) Max. 0.02 ppm
Antimony (Sb) Max. 0.01 ppm
Tin (Sn) Max. 0.02 ppm
Strontium (Sr) Max. 0.01 ppm
Titanium (Ti) Max. 0.01 ppm
Thallium (Tl) Max. 0.01 ppm
Vanadium (V) Max. 0.01 ppm
Zinc (Zn) Max. 0.02 ppm
Zirconium (Zr) Max. 0.01 ppm
Chloride (Cl) Max. 0.1 ppm
Sulfate (SO₄) Max. 1 ppm
KMnO₄ reducing matter (as O) Max. 2 ppm
Aldehydes (as HCHO) Max. 10 ppm
Ethanol (GC) Max. 100 ppm
Methanol (GC) Max. 500 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR