Hydrochloric acid fuming 37%, Semiconductor grade (Honeywell 17823)
Supplier: Honeywell Research Chemicals
|
Danger
|
40253H-2.5L
BJ40253H-2.5LEA
156.27
USD
BJ40253H-2.5L
Hydrochloric acid fuming 37%, Semiconductor grade (Honeywell 17823)
Hydrochloric acid
We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
- Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
- Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Beilstein Number: 1098214
Formula:
HCl Boiling Pt: 42 °C (1.013 hPa) Melting Pt: –35 °C Density: 1.190 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011324 CAS Number: 7647-01-0 UN: 1789 ADR: 8,II |
Specification Test Results
Assay | 37 - 38 % |
Residue on ignition (SO₄) | Max. 5 ppm |
Ammonium (NH₄) | Max. 2 ppm |
Silver (Ag) | Max. 0.02 ppm |
Aluminium (Al) | Max. 0.05 ppm |
Arsenic (As) | Max. 0.01 ppm |
Gold (Au) | Max. 0.02 ppm |
Boron (B) | Max. 0.02 ppm |
Barium (Ba) | Max. 0.1 ppm |
Beryllium (Be) | Max. 0.01 ppm |
Bismuth (Bi) | Max. 0.02 ppm |
Calcium (Ca) | Max. 0.2 ppm |
Cadmium (Cd) | Max. 0.01 ppm |
Cobalt (Co) | Max. 0.01 ppm |
Chromium (Cr) | Max. 0.01 ppm |
Copper (Cu) | Max. 0.01 ppm |
Iron (Fe) | Max. 0.2 ppm |
Gallium (Ga) | Max. 0.02 ppm |
Indium (In) | Max. 0.02 ppm |
Potassium (K) | Max. 0.1 ppm |
Lithium (Li) | Max. 0.02 ppm |
Magnesium (Mg) | Max. 0.1 ppm |
Manganese (Mn) | Max. 0.01 ppm |
Molybdenum (Mo) | Max. 0.01 ppm |
Sodium (Na) | Max. 0.5 ppm |
Nickel (Ni) | Max. 0.01 ppm |
Lead (Pb) | Max. 0.02 ppm |
Platinum (Pt) | Max. 0.02 ppm |
Antimony (Sb) | Max. 0.01 ppm |
Tin (Sn) | Max. 0.02 ppm |
Strontium (Sr) | Max. 0.02 ppm |
Titanium (Ti) | Max. 0.01 ppm |
Thallium (Tl) | Max. 0.02 ppm |
Vanadium (V) | Max. 0.01 ppm |
Zinc (Zn) | Max. 0.05 ppm |
Zirconium (Zr) | Max. 0.01 ppm |
Bromide (Br) | Max. 50 ppm |
Free chlorine (Cl) | Max. 0.5 ppm |
Phosphate (PO₄) | Max. 0.5 ppm |
Sulfate (SO₄) | Max. 1 ppm |
Sulfite (SO₃) | Max. 1 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...