Nitric acid ≥65%, Semiconductor grade (Honeywell 17818)

Supplier: Honeywell Research Chemicals
Danger

40230H-2.5L
BJ40230H-2.5LEA 163.68 USD
BJ40230H-2.5L
Nitric acid ≥65%, Semiconductor grade (Honeywell 17818)
Nitric acid

We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:


  • Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
  • Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.


RTECS Number: 231-714-2

Formula: HNO₃
Boiling Pt: 122 °C (1.013 hPa)
Melting Pt: –31 °C
Density: 1.400…1.480 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8,II

Order Now

Specification Test Results

Assay Min. 65 %
Residue on ignition (SO₄) Max. 5 ppm
Silver (Ag) Max. 0.01 ppm
Aluminium (Al) Max. 0.05 ppm
Arsenic (As) Max. 0.01 ppm
Gold (Au) Max. 0.02 ppm
Boron (B) Max. 0.05 ppm
Barium (Ba) Max. 0.01 ppm
Beryllium (Be) Max. 0.01 ppm
Bismuth (Bi) Max. 0.02 ppm
Calcium (Ca) Max. 0.1 ppm
Cadmium (Cd) Max. 0.01 ppm
Cobalt (Co) Max. 0.01 ppm
Chromium (Cr) Max. 0.01 ppm
Copper (Cu) Max. 0.01 ppm
Iron (Fe) Max. 0.1 ppm
Gallium (Ga) Max. 0.02 ppm
Germanium (Ge) Max. 0.05 ppm
Indium (In) Max. 0.02 ppm
Potassium (K) Max. 0.1 ppm
Lithium (Li) Max. 0.01 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.01 ppm
Molybdenum (Mo) Max. 0.01 ppm
Sodium (Na) Max. 0.3 ppm
Nickel (Ni) Max. 0.02 ppm
Lead (Pb) Max. 0.01 ppm
Platinum (Pt) Max. 0.02 ppm
Antimony (Sb) Max. 0.01 ppm
Tin (Sn) Max. 0.02 ppm
Strontium (Sr) Max. 0.01 ppm
Titanium (Ti) Max. 0.01 ppm
Thallium (Tl) Max. 0.02 ppm
Vanadium (V) Max. 0.01 ppm
Zinc (Zn) Max. 0.05 ppm
Zirconium (Zr) Max. 0.01 ppm
Chloride (Cl) Max. 0.5 ppm
Phosphate (PO₄) Max. 0.5 ppm
Sulfate (SO₄) Max. 0.5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR