Ammonium fluoride solution ≥40%, Semiconductor grade (Honeywell 17767)

Supplier: Honeywell Research Chemicals
Danger

40208H-5L
BJ40208H-5LEA 313.09 USD
BJ40208H-5L
Ammonium fluoride solution ≥40%, Semiconductor grade (Honeywell 17767)
Ammonium fluoride

We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:


  • Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
  • Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.


UNSPSC Code: 12352300

Formula: NH₄F
Boiling Pt: >100 °C (1.013 hPa)
Density: 1.110 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011423
CAS Number: 12125-01-8
UN: 3287
ADR: 6.1,III

Order Now

Specification Test Results

Assay Min. 40 %
Residue on ignition (SO₄) Max. 10 ppm
pH (1 %,20°C) 6.2 - 7.0
Silver (Ag) Max. 0.05 ppm
Aluminium (Al) Max. 0.5 ppm
Arsenic (As) Max. 0.05 ppm
Gold (Au) Max. 0.05 ppm
Barium (Ba) Max. 0.1 ppm
Beryllium (Be) Max. 0.05 ppm
Bismuth (Bi) Max. 0.05 ppm
Calcium (Ca) Max. 0.5 ppm
Cadmium (Cd) Max. 0.1 ppm
Cobalt (Co) Max. 0.05 ppm
Chromium (Cr) Max. 0.1 ppm
Copper (Cu) Max. 0.1 ppm
Iron (Fe) Max. 0.5 ppm
Gallium (Ga) Max. 0.1 ppm
Indium (In) Max. 0.05 ppm
Potassium (K) Max. 0.1 ppm
Lithium (Li) Max. 0.1 ppm
Magnesium (Mg) Max. 0.2 ppm
Manganese (Mn) Max. 0.05 ppm
Molybdenum (Mo) Max. 0.05 ppm
Sodium (Na) Max. 0.2 ppm
Nickel (Ni) Max. 0.05 ppm
Lead (Pb) Max. 0.1 ppm
Platinum (Pt) Max. 0.05 ppm
Antimony (Sb) Max. 0.05 ppm
Silicon (Si) Max. 200 ppm
Tin (Sn) Max. 0.1 ppm
Strontium (Sr) Max. 0.1 ppm
Titanium (Ti) Max. 0.1 ppm
Thallium (Tl) Max. 0.05 ppm
Vanadium (V) Max. 0.1 ppm
Zinc (Zn) Max. 0.1 ppm
Zirconium (Zr) Max. 0.05 ppm
Chloride (Cl) Max. 4 ppm
Phosphate (PO₄) Max. 1 ppm
Sulfate (SO₄) Max. 2 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR