Methanol : Water (50:50)% (v/v), LabReady™, Solvent Blend for HPLC, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger

400-4
BJ400-4CS 2694 USD
BJ400-4 BJNP400-20
Methanol : Water (50:50)% (v/v), LabReady™, Solvent Blend for HPLC, Burdick & Jackson™
Methanol (15 - < 30%)

LabReady™ Solvent Blends


  • Using custom blends not only helps you reduce waste through human error, but also cuts disposal costs for hazardous materials used during blend preparation
  • Honeywell Research Chemicals uses proprietary, closed-loop blending technology, which eliminates variation, ensuring your blends are as precise as possible
  • Honeywell LabReady Blends undergo full safety analyses, with a safety data sheet provided for every blended product
  • Honeywell’s high-precision blending technology reduces contamination and exposure during the blending process, helping to minimize environmental risk

Formula: CH₄O/H₂O
Density: 0.9264 g/cm³ (25 °C)
Flash Pt: 29 °C
Storage Temperature: Ambient
MDL Number: MFCD00004595
EINECS: 200-659-6
UN: 1230
ADR: 3,II

Order Now

Specification Test Results

Water (v/v) 49.0 - 51.0%
Methanol (v/v) 49.0 - 51.0%
UV Cutoff 198 nm
Residue < 1 mg/L
Color (APHA) < 10
HPLC Gradient @ 220 nm < 0.005 AU
HPLC Gradient @ 254 nm < 0.001 AU

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR