Sulfuric acid concentrate for dilution to 1L, 0.005 M H2SO4 (0.01N) volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals
Warning

38308-1EA
BJ38308-1EAEA 103.94 USD
BJ38308-1EA
Sulfuric acid concentrate for dilution to 1L, 0.005 M H2SO4 (0.01N) volumetric solution, Fluka™
Sulfuric acid

concentrate for 1L standard solution

Formula: H₂SO₄
Boiling Pt: 100 °C (1.013 hPa)
Density: 1.010 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
UN: 3264
ADR: 8,III

Order Now

Specification Test Results

Factor 0.998 - 1.002
BAM/EMPA reference material effective
BAM/EMPA Certificate Number effective
Traceable to NIST SRM effective


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR