Sulfuric acid concentrate for dilution to 1 L, 1 M H2SO4 (2.0 N) volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals
Danger

38291-1EA
BJ38291-1EAEA 97.99 USD
BJ38291-1EA
Sulfuric acid concentrate for dilution to 1 L, 1 M H2SO4 (2.0 N) volumetric solution, Fluka™
Sulfuric acid

concentrate for 1L standard solution

Formula: H₂SO₄
Boiling Pt: >100 °C (1.013 hPa)
Density: 1.180 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
UN: 2796
ADR: 8,II

Order Now

Specification Test Results

Factor 0.998 - 1.002
BAM/EMPA reference material effective
BAM/EMPA Certificate Number effective
Traceable to NIST SRM effective


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR