Hydrochloric acid concentrate for dilution to 2L, 1 M HCl (1N) standard volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals
Danger

38281-1EA
BJ38281-1EAEA 97.09 USD
BJ38281-1EA
Hydrochloric acid concentrate for dilution to 2L, 1 M HCl (1N) standard volumetric solution, Fluka™
Hydrochloric acid

concentrate for 2L standard solution

Formula: HCl
Boiling Pt: 100 °C
Melting Pt: 0 °C
Density: 1.11 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8,II

Order Now

Specification Test Results

Factor 0.998 - 1.002
BAM/EMPA reference material effective
BAM/EMPA Certificate Number effective
Traceable to NIST SRM effective


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR