Oxalic acid concentrate for dilution to 1L, 0.005 M (COOH)2 (0.01N) volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals

38255-1EA
BJ38255-1EAEA 69 USD
BJ38255-1EA
Oxalic acid concentrate for dilution to 1L, 0.005 M (COOH)2 (0.01N) volumetric solution, Fluka™
Oxalic acid
Formula: C₂H₂O₄
Storage Temperature: Ambient
MDL Number: MFCD00002573
CAS Number: 144-62-7
EINECS: 231-791-2

Order Now

Specification Test Results

Factor 0.998 - 1.002


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR