Sodium hydroxide concentrate for dilution to 1 L, 0.01 M NaOH (0.01 N) volumetric solution, Fluka™
Supplier: Honeywell Research Chemicals
|
Warning
|
Synonyms:
Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye
38227-1EA
38227-6X1EA
BJ38227-1EAEA
75.64
USD
BJ38227-1EA
BJ38227-6X1EA
Sodium hydroxide concentrate for dilution to 1 L, 0.01 M NaOH (0.01 N) volumetric solution, Fluka™
Sodium hydroxide
For 1L standard solution
Formula:
NaOH Boiling Pt: 100 °C (1.013 hPa) Melting Pt: no data available Density: 1.000 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 UN: 1824 ADR: 8,III |
Specification Test Results
Factor | 0.998 - 1.002 |
BAM/EMPA reference material | effective |
BAM/EMPA Certificate Number | effective |
Traceable to NIST SRM | effective |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...