Sodium hydroxide concentrate for dilution to 1 L, 1.0 M NaOH (1.0 N) volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals
Danger

38215-1EA
BJ38215-1EAEA 62.46 USD
BJ38215-1EA
Sodium hydroxide concentrate for dilution to 1 L, 1.0 M NaOH (1.0 N) volumetric solution, Fluka™
Sodium hydroxide

concentrate for 1L standard solution

Formula: NaOH
Boiling Pt: 120 °C
Melting Pt: 319 °C
Density: 1.350 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

Factor 0.998 - 1.002
BAM/EMPA reference material effective
BAM/EMPA Certificate Number effective
Traceable to NIST SRM effective


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR