Magnesium sulfate concentrate for dilution to 1L, 0.1 M MgSO4 volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals

Synonyms: MgSO4, Magnesium sulfate anhydrous, Magnesium sulphate anhydrous

38146-1EA
BJ38146-1EAEA 119.94 USD
BJ38146-1EA
Magnesium sulfate concentrate for dilution to 1L, 0.1 M MgSO4 volumetric solution, Fluka™
Magnesium sulphate
Formula: MgSO₄
MW: 120.37 g/mol
Melting Pt: 1127
Storage Temperature: Ambient
MDL Number: MFCD00011110
CAS Number: 7487-88-9
Merck Index: 12,05731
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bc2ae189b0cbc1bfc599951528466503d0d9008&vid=63867221020ebd2c8c227492e609a4764639ff79&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Factor 0.998 - 1.002


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR