Potassium thiocyanate concentrate for dilution to 1L, 0.1 M KSCN volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals
Warning

38140-1EA 38140-6X1EA
BJ38140-1EAEA 64.83 USD
BJ38140-1EA BJ38140-6X1EA
Potassium thiocyanate concentrate for dilution to 1L, 0.1 M KSCN volumetric solution, Fluka™
Potassium thiocyanate
Formula: KSCN
Density: 1 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011413
CAS Number: 333-20-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR