Potassium iodate concentrate for dilution to 1L, 1/60 M KIO3 (0.1N) volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals

38120-1EA
BJ38120-1EAEA 85.46 USD
BJ38120-1EA
Potassium iodate concentrate for dilution to 1L, 1/60 M KIO3 (0.1N) volumetric solution, Fluka™
Potassium iodate
Formula: KlO₃
MW: 214 g/mol
Boiling Pt: 100 °C
Melting Pt: 560
Storage Temperature: Ambient
MDL Number: MFCD00011406
CAS Number: 7758-05-6
Merck Index: 13,07726

Order Now

Specification Test Results

Factor 0.998 - 1.002


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR