Potassium dichromate concentrate for dilution to 1L, 1/60 M K2Cr2O7 (0.1N) volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals
Danger

38100-1EA
BJ38100-1EAEA 94.35 USD
BJ38100-1EA BJ38100-6X1EA
Potassium dichromate concentrate for dilution to 1L, 1/60 M K2Cr2O7 (0.1N) volumetric solution, Fluka™
Potassium dichromate

For 1L standard solution

Formula: K₂Cr₂O₇
Boiling Pt: 100 °C (1.013 hPa)
Melting Pt: no data available
Density: 1.030 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011367
CAS Number: 7778-50-9
UN: 2922
ADR: 8,II

Order Now

Specification Test Results

Factor 0.998 - 1.002


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR