Potassium bromate concentrate for dilution to 1L, 1/60 M KBrO3 (0.1N) volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals
Danger

38080-1EA
BJ38080-1EAEA 80.89 USD
BJ38080-1EA
Potassium bromate concentrate for dilution to 1L, 1/60 M KBrO3 (0.1N) volumetric solution, Fluka™
Potassium bromate
Formula: KBrO₃
MW: 167 g/mol
Density: 1.024 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011359
CAS Number: 7758-01-2

Order Now

Specification Test Results

Factor 0.998 - 1.002

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR