Iodine concentrate for dilution to 1L, 0.05 M I2 (0.1 N) volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals
Danger

38060-6X1EA
BJ38060-1EACS 378.15 USD
BJ38060-1EA
Iodine concentrate for dilution to 1L, 0.05 M I2 (0.1 N) volumetric solution, Fluka™
Iodine

concentrate for 1L standard solution

Formula: I₂
Melting Pt: 113.5 °C
Density: 4.930 g/cm3
Storage Temperature: Ambient
MDL Number: MFCD00011355
CAS Number: 7553-56-2
UN: 3495
ADR: 8,III

Order Now

Specification Test Results

Factor 0.998 - 1.002


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR