Acetic acid concentrate for dilution to 1L, 1.0 M CH3COOH (1N) volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals
Danger

38051-1EA
BJ38051-1EAEA 106.5 USD
BJ38051-1EA BJ38051-6X1EA
Acetic acid concentrate for dilution to 1L, 1.0 M CH3COOH (1N) volumetric solution, Fluka™
Acetic acid

For 1L standard solution

Formula: H₃CCOOH
Boiling Pt: 117…118 °C
Melting Pt: 4 °C
Flash Pt: 19.9 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
UN: 2789
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Factor 0.998 - 1.002


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR