Acetic acid concentrate for dilution to 1L, 0.1 M CH3COOH (0.1N) volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals
Danger

38050-1EA
BJ38050-1EAEA 123.29 USD
BJ38050-1EA
Acetic acid concentrate for dilution to 1L, 0.1 M CH3COOH (0.1N) volumetric solution, Fluka™
Acetic acid

concentrate for 1L standard solution

Formula: H₃CCOOH
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
UN: 2790
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Factor 0.998 - 1.002


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR