1-Butanol ≥99.4% ACS, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Propylcarbinol, Methylolpropane, Butan-1-ol, Butyl alcohol, n-Butyl alcohol, Butyl hydroxide, 1-Butyl alcohol, Butyl Alcohol, 1-Hydroxybutane, 1-Hydroxy butane, n-Butan-1-ol, Propyl carbinol, n-Butanol, n-Butyl Alcohol

360465-2.5L 360465-4X2.5L
BJ360465-2.5LEA 227 USD
BJ360465-2.5L BJ3604654X2.5L
1-Butanol ≥99.4% ACS, Riedel-de Haën™
1-Butanol

ACS GENERAL USE SOLVENTS


  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications


Beilstein Number: 969148
RTECS Number: EO1400000
UNSPSC Code: 12191601

Formula: H₃C(CH₂)₂CH₂OH
MW: 74.12 g/mol
Boiling Pt: 117.6
Melting Pt: -89
Density: 0.81 g/cm³ (20 °C)
Flash Pt: 9.4 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00002964
CAS Number: 71-36-3
EINECS: 200-751-6
UN: 1120
ADR: 3 III
Merck Index: 13,01539
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9395eeb6150a74222da52460f53cedeccd93442&vid=fd8352a556914c5be7213c576355a88e130d1a7b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Autoignition Temperature 649 °F
Identity (IR) complying
Assay (GC) Min. 99.40 %
Non-volatile matter Max. 0.005 %
Water (Karl Fischer) Max. 0.1 %
Titrable Acid Max. 0.0008 meq/g
Di-n-butyl ether (GC) Max. 0.2 %
n-Butyraldehyde (GC) Max. 0.01 %
APHA Max. 10
Explosion Limit 11.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR