Tetramethylammonium hydroxide 0.1 M (CH3)4NOH (0.1N) in 2-propanol/methanol volumetric solution, for titration in non-aqueous media, Fluka™

Supplier: Honeywell Research Chemicals
Danger

35436-1L
BJ35436-1LEA 357.27 USD
BJ35436-1L
Tetramethylammonium hydroxide 0.1 M (CH3)4NOH (0.1N) in 2-propanol/methanol volumetric solution, for titration in non-aqueous media, Fluka™
Tetramethylammonium hydroxide
Formula: C₄H₁₃NO
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00008280
CAS Number: 75-59-2
UN: 1992
ADR: 3,II

Order Now

Specification Test Results

Factor 0.999 - 1.001


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR