Zinc sulfate 0.1 M ZnSO4 ≥99.5% volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals
Warning

35392-1L
BJ35392-1LEA 106 USD
BJ35392-1L
Zinc sulfate 0.1 M ZnSO4 ≥99.5% volumetric solution, Fluka™
Zinc sulphate heptahydrate
Formula: ZnSO₄·7H₂O
Storage Temperature: Ambient
MDL Number: MFCD00149894
CAS Number: 7446-20-0
ADR: 9,III

Order Now

Specification Test Results

Factor 0.999 - 1.001


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR