Sulfuric acid 0.1 M H2SO4 (0.2N) in aqueous solution Reag. Ph. Eur. volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals
Warning

35357-6X1L 35357-5L-VP
BJ35357-1LCS 491.58 USD
BJ35357-1L BJ35357-5L-VP
Sulfuric acid 0.1 M H2SO4 (0.2N) in aqueous solution Reag. Ph. Eur. volumetric solution, Fluka™
Sulfuric acid
Formula: H₂SO₄
MW: 98.08
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.010 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 203-755-4
UN: 3264
ADR: 8 III

Order Now

Specification Test Results

Factor 0.999 - 1.001
BAM/EMPA reference material effective
BAM/EMPA Certificate Number effective
Traceable to NIST SRM effective


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR