Sulfuric acid 0.25 M H2SO4 (0.5N) in aqueous solution ≥99.9% Reag. Ph. Eur. volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals
Warning

35355-10L-VP 35355-1L
BJ35355-10L-VPEA 300.16 USD
BJ35355-10L-VP BJ-35355-1L
Sulfuric acid 0.25 M H2SO4 (0.5N) in aqueous solution ≥99.9% Reag. Ph. Eur. volumetric solution, Fluka™
Sulfuric acid
Formula: H₂SO₄
MW: 98.08
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.020 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 203-755-4
UN: 3264
ADR: 8 III

Order Now

Specification Test Results

Factor 0.999 - 1.001
BAM/EMPA reference material effective
BAM/EMPA Certificate Number effective
Traceable to NIST SRM effective


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR