Sulfuric acid 0.5 M H2SO4 (1.0N) in aqueous solution ≥99.9% Reag. Ph. Eur. volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals
Danger

35354-10L-VP 35354-6X1L 35354-5L-VP 35354-2.5L
BJ35354-10L-VPEA 217.54 USD
BJ35354-10L-VP BJ35354-1L BJ35354-5L-VP BJ-35354-2.5L BJ-35354-5L
Sulfuric acid 0.5 M H2SO4 (1.0N) in aqueous solution ≥99.9% Reag. Ph. Eur. volumetric solution, Fluka™
Sulfuric acid
Formula: H₂SO₄
Boiling Pt: 100 °C (1.013 hPa)
Melting Pt: <0 °C
Density: 1.030 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 203-755-4
UN: 3264
ADR: 8,III

Order Now

Specification Test Results

Factor 0.999 - 1.001
BAM/EMPA reference material effective
BAM/EMPA Certificate Number effective
Traceable to NIST SRM effective


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR