Nitric acid 0.1 M HNO3 (0.1N) volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals
Warning

35318-1L
BJ35318-1LEA 153.07 USD
BJ35318-1L
Nitric acid 0.1 M HNO3 (0.1N) volumetric solution, Fluka™
Nitric acid
Formula: HNO₃
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-791-2
UN: 3264
ADR: 8,III

Order Now

Specification Test Results

Factor 0.999 - 1.001
BAM/EMPA reference material effective
BAM/EMPA Certificate Number effective
Traceable to NIST SRM effective


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR