Trifluoromethanesulfonic acid 0.1 M CF3SO3H (0.1 N) in acetic acid volumetric solution, for titration in non-aqueous media, Fluka™

Supplier: Honeywell Research Chemicals
Danger

35317-1L
BJ35317-1LEA 214.28 USD
BJ35317-1L
Trifluoromethanesulfonic acid 0.1 M CF3SO3H (0.1 N) in acetic acid volumetric solution, for titration in non-aqueous media, Fluka™
Trifluoromethanesulphonic acid
Formula: CHF₃O₃S
Boiling Pt: 117 °C (1.013 hPa)
Density: 1.059 g/cm³ (20 °C)
Flash Pt: 38 °C
Storage Temperature: Ambient
MDL Number: MFCD00007514
CAS Number: 1493-13-6
EINECS: 216-087-5
UN: 2789
ADR: 8,II

Order Now

Specification Test Results

Factor 0.999 - 1.001
BAM/EMPA reference material effective
BAM/EMPA Certificate Number effective
Traceable to NIST SRM effective


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR