Sodium thiosulfate 0.2 M Na2S2O3 (0.2N) volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals

35233-6X1L
BJ35233-1LCS 927.35 USD
BJ35233-1L BJ35233-5L-VP
Sodium thiosulfate 0.2 M Na2S2O3 (0.2N) volumetric solution, Fluka™
Sodium thiosulphate
Formula: Na₂S₂O₃
Boiling Pt: 100 °C
Density: 1.01 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003499
CAS Number: 7772-98-7

Order Now

Specification Test Results

Factor 0.998 - 1.002


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR