Sodium acetate 0.1 M CH3COONa in acetic acid volumetric solution, Fluka™
Supplier: Honeywell Research Chemicals
35208-1L
BJ35208-1LEA
182.59
USD
BJ35208-1L
Sodium acetate 0.1 M CH3COONa in acetic acid volumetric solution, Fluka™
Sodium acetate
Honeywell's high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Beilstein Number: 3786729
Specification Test Results
Factor | 0.99 - 1.01 |