Potassium thiocyanate 0.1 M KSCN volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals

Synonyms: Potassium rhodanide

35194-1L
BJ35194-1LEA 153.93 USD
BJ35194-1L
Potassium thiocyanate 0.1 M KSCN volumetric solution, Fluka™
Potassium thiocyanate
Formula: KSCN
Density: 1.886 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011413
CAS Number: 333-20-0
EINECS: 231-791-2

Order Now

Specification Test Results

Factor 0.999 - 1.001


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR