Potassium permanganate 0.02 M KMnO4 (0.1N) volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals

35186-6X1L
BJ35186-1LCS 369.95 USD
BJ35186-1L
Potassium permanganate 0.02 M KMnO4 (0.1N) volumetric solution, Fluka™
Potassium permanganate
Formula: KMnO₄
Storage Temperature: Ambient
MDL Number: MFCD00011364
CAS Number: 7722-64-7
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Factor 0.999 - 1.001
BAM/EMPA reference material effective
BAM/EMPA Certificate Number effective
Traceable to NIST SRM effective


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR