Copper(II) sulfate 0.1 M CuSO4 volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals

Synonyms: Cupric sulphate, Cupric sulfate

35185-1L
BJ35185-1LEA 240 USD
BJ35185-1L
Copper(II) sulfate 0.1 M CuSO4 volumetric solution, Fluka™
Copper (II) sulphate
Formula: CuSO₄
MW: 159.61 g/mol
Boiling Pt: >100 °C (1.013 hPa)
Density: 1.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010981
CAS Number: 7758-98-7

Order Now

Specification Test Results

Factor 0.999 - 1.001

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR